missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Synaptotagmin-1 Protein

A DNA sequence encoding the Homo sapiens (Human) Synaptotagmin-1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6757-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16095600

  • 530.00 € / 100µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001129277.1
Synaptotagmin-1 Protein
Research Use Only
SYT1
Recombinant Protein
E. coli
KNAINMKDVK DLGKTMKDQA LKDDDAETGL TDGEEKEEPK EEEKLGKLQY SLDYDFQNNQ LLVGIIQAAE LPALDMGGTS DPYVKVFLLP DKKKKFETKV HRKTLNPVFN EQFTFKVPYS ELGGKTLVMA VYDFDRFSKH DIIGEFKVPM NTVDFGHVTE EWRDLQSAEK EEQEKLGDIC FSLRYVPTAG KLTVVILEAK NLKKMDVGGL SDPYVKIHLM QNGKRLKKKK TTIKKNTLNP YYNESFSFEV PFEQIQKVQV VVTVLDYDKI GKNDAIGKVF VGYNSTGAEL RHWSDMLANP RRPIAQWHTL QVEEEVDA
Greater than 90% as determined by SDS-PAGE.
6857
100 μg
Not determined.
Cytoplasmic Domain
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions