missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUCLG2 Protein
A DNA sequence encoding the Homo sapiens (Human) SUCLG2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7942-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001171070.1 | |
SUCLG2 Protein | |
Research Use Only | |
SUCLG2 | |
Recombinant Protein | |
E. coli | |
MSDNGVRVQR FFVADTANEA LEAAKRLNAK EIVLKAQILA GGRGKGVFNS GLKGGVHLTK DPNVVGQLAK QMIGYNLATK QTPKEGVKVN KVMVAEALDI SRETYLAILM DRSCNGPVLV GSPQGGVDIE EVAASNPELI FKEQIDIFEG IKDSQAQRMA ENLGFVGPLK SQAADQITKL YNLFLKIDAT QVEVNPFGET PEGQVVCFDA KINFDDNAEF RQKDIFAMDD KSENEPIENE AAKYDLKYIG LDGNIACFVN GAGLAMATCD IIFLNGGKPA NFLDLGGGVK EAQVYQAFKL LTADPKVEAI LVNIFGGIVN CAIIANGITK ACRELELKVP LVVRLEGTNV QEAQKILNNS GLPITSAIDL EDAAK | |
Greater than 90% as determined by SDS-PAGE. |
8801 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |