missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Solute carrier family 41 member 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Solute carrier family 41 member 2, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_115524.3 |
---|---|
Gene ID (Entrez) | 84102 |
Name | Solute carrier family 41 member 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035180
|
enQuireBio™
QP6695-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055180
|
enQuireBio™
QP6695-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045180
|
enQuireBio™
QP6695-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065180
|
enQuireBio™
QP6695-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075180
|
enQuireBio™
QP6695-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085180
|
enQuireBio™
QP6695-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_115524.3 | |
Solute carrier family 41 member 2 Protein | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
84102 | |
Research Use Only | |
SLC41A2 | |
Recombinant Protein | |
E. coli | |
MTNSKGRSIT DKTSGGPSSG GGFVDWTLRL NTIQSDKFLN LLLSMVPVIY QKNQEDRHKK ANGIWQDGLS TAVQTFSNRS EQHMEYHSFS EQSFHANNGH ASSSCSQKYD DYANYNYCDG RETSETTAML QDEDISSDGD EDAIVEVTPK LPKESSGIMA LQ | |
Greater than 90% as determined by SDS-PAGE. |