missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SMARCB1 Protein

A DNA sequence encoding the Homo sapiens (Human) SMARCB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7605-ba-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16079700

Quantity Price
1 690.89 € / 20µg
5 648.34 € / 20µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001007469.1
SMARCB1 Protein
20 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
6598
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SMARCB1
Recombinant Protein
Baculovirus
MMMALSKTFG QKPVKFQLED DGEFYMIGSE VGNYLRMFRG SLYKRYPSLW RRLATVEERK KIVASSHGKK TKPNTKDHGY TTLATSVTLL KASEVEEILD GNDEKYKAVS ISTEPPTYLR EQKAKRNSQW VPTLPNSSHH LDAVPCSTTI NRNRMGRDKK RTFPLCFDDH DPAVIHENAS QPEVLVPIRL DMEIDGQKLR DAFTWNMNEK LMTPEMFSEI LCDDLDLNPL TFVPAIASAI RQQIESYPTD SILEDQSDQR VIIKLNIHVG NISLVDQFEW DMSEKENSPE KFALKLCSEL GLGGEFVTTI AYSIRGQLSW HQKTYAFSEN PLPTVEIAIR NTGDADQWCP LLETLTDAEM EKKIRDQDRN TRRMR
Videos
SDS
Documents
Certificates
Promotions

Promotions