missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Signal recognition particle 19 kDa Protein

A DNA sequence encoding the Homo sapiens (Human) Signal recognition particle 19 kDa protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6728-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16055420

  • 2385.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001191122.1
Signal recognition particle 19 kDa Protein
Research Use Only
SRP19
Recombinant Protein
E. coli
ACAAARSPAD QDRFICIYPA YLNNKKTIAE GRRIPISKAV ENPTATEIQD VCSAVGLNVF LEKNKMYSRE WNRDVQYRGR VRVQLKQEDG SLCLVQFPSR KSVMLYAAEM IPKLKTRTQK TGGADQSLQQ GEGSKKGKGK KKK
Greater than 90% as determined by SDS-PAGE.
6728
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions