missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SerpinA6 / CBG Protein

A DNA sequence encoding the Homo sapiens (Human) SerpinA6 / CBG, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6667-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084940

Quantity Price
1 2248.90 € / 1mg
3 2115.77 € / 1mg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001747.2
SerpinA6 / CBG Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
866
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SERPINA6
Recombinant Protein
E. coli
MDPNAAYVNM SNHHRGLASA NVDFAFSLYK HLVALSPKKN IFISPVSISM ALAMLSLGTC GHTRAQLLQG LGFNLTERSE TEIHQGFQHL HQLFAKSDTS LEMTMGNALF LDGSLELLES FSADIKHYYE SEVLAMNFQD WATASRQINS YVKNKTQGKI VDLFSGLDSP AILVLVNYIF FKGTWTQPFD LASTREENFY VDETTVVKVP MMLQSSTISY LHDSELPCQL VQMNYVGNGT VFFILPDKGK MNTVIAALSR DTINRWSAGL TSSQVDLYIP KVTISGVYDL GDVLEEMGIA DLFTNQANFS RITQDAQLKS SKVVHKAVLQ LNEEGVDTAG STGVTLNLTS KPIILRFNQP FIIMIFDHFT WSSLFLARVM NPV
Videos
SDS
Documents
Certificates
Promotions

Promotions