missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Serine / threonine-protein kinase PAK 4 Protein
A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase PAK 4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6464-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001014831.1 | |
Serine / threonine-protein kinase PAK 4 Protein | |
Research Use Only | |
PAK4 | |
Recombinant Protein | |
E. coli | |
MFGKRKKRVE ISAPSNFEHR VHTGFDQHEQ KFTGLPRQWQ SLIEESARRP KPLVDPACIT SIQPGAPKTI VRGSKGAKDG ALTLLLDEFE NMSVTRSNSL RRDSPPPPAR ARQENGMPEK PPGPRSPQRE PQRVSHEQFR AALQLVVDPG DPRSYLDNFI KIGEGSTGIV CIATVRSSGK LVAVKKMDLR KQQRRELLFN EVVIMRDYQH ENVVEMYNSY LVGDELWVVM EFLEGGALTD IVTHTRMNEE QIAAVCLAVL QALSVLHAQG VIHRDIKSDS ILLTHDGRVK LSDFGFCAQV SKEVPRRKSL VGTPYWMAPE LISRLPYGPE VDIWSLGIMV IEMVDGEPPY FNEPPLKAMK MIRDNLPPRL KNLHKVSPSL KGFLDRLLVR DPAQRATAAE LLKHPFLAKA GPPASIVPLM RQNRTR | |
Greater than 90% as determined by SDS-PAGE. |
10298 | |
100 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |