missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serine / threonine-protein kinase PAK 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase PAK 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7527-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16099180

  • 2385.00 € / 1mg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001122092.1
Serine / threonine-protein kinase PAK 1 Protein
Research Use Only
PAK1
Recombinant Protein
E. coli
MSNNGLDIQD KPPAPPMRNT STMIGAGSKD AGTLNHGSKP LPPNPEEKKK KDRFYRSILP GDKTNKKKEK ERPEISLPSD FEHTIHVGFD AVTGEFTGMP EQWARLLQTS NITKSEQKKN PQAVLDVLEF YNSKKTSNSQ KYMSFTDKSA EDYNSSNALN VKAVSETPAV PPVSEDEDDD DDDATPPPVI APRPEHTKSV YTRSVIEPLP VTPTRDVATS PISPTENNTT PPDALTRNTE KQKKKPKMSD EEILEKLRSI VSVGDPKKKY TRFEKIGQGA SGTVYTAMDV ATGQEVAIKQ MNLQQQPKKE LIINEILVMR ENKNPNIVNY LDSYLVGDEL WVVMEYLAGG SLTDVVTETC MDEGQIAAVC RECLQALEFL HSNQVIHRDI KSDNILLGMD GSVKLTDFGF CAQITPEQSK RSTMVGTPYW MAPEVVTRKA YGPKVDIWSL GIMAIEMIEG EPPYLNENPL RALYLIATNG TPELQNPEKL SAIFRDFLNR CLEMDVEKRG SAKELLQHQF LKIAKPLSSL TPLIAAAKEA TKNNH
Greater than 90% as determined by SDS-PAGE.
5058
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions