missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Serine / threonine-protein kinase NIM1 Protein

A DNA sequence encoding the Homo sapiens (Human) Serine/threonine-protein kinase NIM1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7811-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16070941

  • 493.03 € / 100µg

Estimated Shipment: 12-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_699192.1
Serine / threonine-protein kinase NIM1 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
167359
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NIM1
Recombinant Protein
E. coli
MTAVYMNGGG LVNPHYARWD RRDSVESGCQ TESSKEGEEG QPRQLTPFEK LTQDMSQDEK VVREITLGKR IGFYRIRGEI GSGNFSQVKL GIHSLTKEKV AIKILDKTKL DQKTQRLLSR EISSMEKLHH PNIIRLYEVV ETLSKLHLVM EYAGGGELFG KISTEGKLSE PESKLIFSQI VSAVKHMHEN QIIHRDLKAE NVFYTSNTCV KVGDFGFSTV SKKGEMLNTF CGSPPYAAPE LFRDEHYIGI YVDIWALGVL LYFMVTGTMP FRAETVAKLK KSILEGTYSV PPHVSEPCHR LIRGVLQQIP TERYGIDCIM NDEWMQGVPY PTPLEPFQLD PKHLSETSTL KEEENEVKST LEHLGITEEH IRNNQGRDAR SSITGVYRII LHRVQRKKAL ESVPVMMLPD PKERDLKKGS RVYRGIRHTS KFCSIL
Videos
SDS
Documents
Certificates
Promotions

Promotions