missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Secretory phospholipase A2 receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Secretory phospholipase A2 receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7606-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16079720

Quantity Price
1 1600.18 € / 100µg
3 1504.97 € / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_001007268.1
Secretory phospholipase A2 receptor Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
22925
Greater than 90% as determined by SDS-PAGE.
Research Use Only
PLA2R1
Recombinant Protein
Mammal
EEKTWHEALR SCQADNSALI DITSLAEVEF LVTLLGDENA SETWIGLSSN KIPVSFEWSN DSSVIFTNWH TLEPHIFPNR SQLCVSAEQS EGHWKVKNCE ERLFYICKKA GHVLSDAESG CQEGWERHGG FCYKID
Videos
SDS
Documents
Certificates
Promotions

Promotions