missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SART3 Protein

A DNA sequence encoding the Homo sapiens (Human) SART3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7616-ye-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16099810

Quantity Price
1 2300.65 € / 1mg
3 2163.76 € / 1mg
Estimated Shipment
09-06-2021
Add to basket


Description

Description

Specifications

Specifications

NP_055521.1
SART3 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
9733
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SART3
Recombinant Protein
Yeast
QRKRARAEKK ALKKKKKIRG PEKRGADEDD EKEWGDDEEE QPSKRRRVEN SIPAAGETQN VEVAAGPAGK CAAVDVEPPS KQKEKAASLK RDMPKVLHDS SKDSITVFVS NLPYSMQEPD TKLRPLFEAC GEVVQIRPIF SNRGDFRGYC YVEFKEEKSA LQALEMDRKS VEGRPMFVSP CVDKSKNPDF KVFRYSTSLE KHKLFISGLP FSCTKEELEE ICKAHGTVKD LRLVTNRAGK PKGLAYVEYE NESQASQAVM KMDGMTIKEN IIKVAISNPP QRKVPEKPET RKAPGGPMLL P
Videos
SDS
Documents