missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A14 / S114 Protein

A DNA sequence encoding the Homo sapiens (Human) S100A14 / S114, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8145-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16092891

  • 2385.00 € / 1mg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_065723.1
S100A14 / S114 Protein
Research Use Only
S100A14
Recombinant Protein
E. coli
MGQCRSANAE DAQEFSDVER AIETLIKNFH QYSVEGGKET LTPSELRDLV TQQLPHLMPS NCGLEEKIAN LGSCNDSKLE FRSFWELIGE AAKSVKLERP VRGH
Greater than 80% as determined by SDS-PAGE.
57402
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions