missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ripk3 Protein

A DNA sequence encoding the Homo sapiens (Human) Ripk3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9945-ba-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16079051

Quantity Price
1 614.12 € / 20µg
5 575.74 € / 20µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_006862.2
Ripk3 Protein
20 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
11035
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Ripk3
Recombinant Protein
Baculovirus
MSCVKLWPSG APAPLVSIEE LENQELVGKG GFGTVFRAQH RKWGYDVAVK IVNSKAISRE VKAMASLDNE FVLRLEGVIE KVNWDQDPKP ALVTKFMENG SLSGLLQSQC PRPWPLLCRL LKEVVLGMFY LHDQNPVLLH RDLKPSNVLL DPELHVKLAD FGLSTFQGGS QSGTGSGEPG GTLGYLAPEL FVNVNRKAST ASDVYSFGIL MWAVLAGREV ELPTEPSLVY EAVCNRQNRP SLAELPQAGP ETPGLEGLKE LMQLCWSSEP KDRPSFQECL PKTDEVFQMV ENNMNAAVST VKDFLSQLRS SNRRFSIPES GQGGTEMDGF RRTIENQHSR NDVMVSEWLN KLNLEEPPSS VPKKCPSLTK RSRAQEEQVP QAWTAGTSSD SMAQPPQTPE TSTFRNQMPS PTSTGTPSPG PRGNQGAERQ GMNWSCRTPE PNPVTGRPLV NIYNCSGVQV GDNNYLTMQQ TTALPTWGLA PSGKGRGLQH PPPVGSQEGP KDPEAWSRPQ GWYNHSGK
Videos
SDS
Documents
Certificates
Promotions

Promotions