missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ribonuclease P protein subunit p20 Protein

A DNA sequence encoding the Homo sapiens (Human) Ribonuclease P protein subunit p20, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6515-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16083840

  • 530.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005828.2
Ribonuclease P protein subunit p20 Protein
Research Use Only
POP7
Recombinant Protein
E. coli
MAENREPRGA VEAELDPVEY TLRKRLPSRL PRRPNDIYVN MKTDFKAQLA RCQKLLDGGA RGQNACSEIY IHGLGLAINR AINIALQLQA GSFGSLQVAA NTSTVELVDE LEPETDTREP LTRIRNNSAI HIRVFRVTPK
Greater than 80% as determined by SDS-PAGE.
10248
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions