missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Regulator of G-protein signaling 10 Protein

A DNA sequence encoding the Homo sapiens (Human) Regulator of G-protein signaling 10, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6602-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084470

  • 530.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001005339.1
Regulator of G-protein signaling 10 Protein
Research Use Only
RGS10
Recombinant Protein
E. coli
MFNRAVSRLS RKRPPSDIHD SDGSSSSSHQ SLKSTAKWAA SLENLLEDPE GVKRFREFLK KEFSEENVLF WLACEDFKKM QDKTQMQEKA KEIYMTFLSS KASSQVNVEG QSRLNEKILE EPHPLMFQKL QDQIFNLMKY DSYSRFLKSD LFLKHKRTEE EEEDLPDAQT AAKRASRIYN T
Greater than 80% as determined by SDS-PAGE.
6001
100 μg
Not determined.
Full Length of Isoform 3
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions