missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBMY1F Protein
A DNA sequence encoding the Homo sapiens (Human) RBMY1F, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001006117.2 |
---|---|
Gene ID (Entrez) | 159163 |
Name | RBMY1F Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16089280
|
enQuireBio™
QP7543-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16019290
|
enQuireBio™
QP7543-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099280
|
enQuireBio™
QP7543-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16009290
|
enQuireBio™
QP7543-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079280
|
enQuireBio™
QP7543-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001006117.2 | |
RBMY1F Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
159163 | |
Research Use Only | |
RBMY1F | |
Recombinant Protein | |
E. coli | |
MVEADHPGKL FIGGLNRETN EKMLKAVFGK HGPISEVLLI KDRTSKSRGF AFITFENPAD AKNAAKDMNG TSLHGKAIKV EQAKKPSFQS GGRRRPPASS RNRSPSGSLR SARGSSGGTR GWLPSHEGHL DDGGYTPDLK MSYSRGLIPV KRGPSSRSGG PPPKKSAPSA VARSNSWMGS QGPMSQRREN YGVPPRRATI SSWRNDRMST RHDGYATNDG NHPSCQETRD YAPPSRGYAY RDNGHSNRDE HSSRGYRNHR SSRETRDYAP PSRGHAYRDY GHSRRDESYS RGYRNHRSSR ETREYAPPSR GHGYRDYGHS RRHESYSRGY RNHPSSRETR DYAPPHRDYA YRDYGHSSWD EHSSRGYSYH DGYGEALGRD HSEHLSGSSY RDALQRYGTS HGAPPARGPR MSYGGSTCHA YSNTRDRYGR SWESYSSCGD FHYCDREHVC RKDQRNPPSL GRVLPDPREA YGSSSYVASI VDGGESRSEK GDSSRY | |
Greater than 90% as determined by SDS-PAGE. |