missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RBMY1F Protein
A DNA sequence encoding the Homo sapiens (Human) RBMY1F, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7543-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001006117.2 | |
RBMY1F Protein | |
Research Use Only | |
RBMY1F | |
Recombinant Protein | |
E. coli | |
MVEADHPGKL FIGGLNRETN EKMLKAVFGK HGPISEVLLI KDRTSKSRGF AFITFENPAD AKNAAKDMNG TSLHGKAIKV EQAKKPSFQS GGRRRPPASS RNRSPSGSLR SARGSSGGTR GWLPSHEGHL DDGGYTPDLK MSYSRGLIPV KRGPSSRSGG PPPKKSAPSA VARSNSWMGS QGPMSQRREN YGVPPRRATI SSWRNDRMST RHDGYATNDG NHPSCQETRD YAPPSRGYAY RDNGHSNRDE HSSRGYRNHR SSRETRDYAP PSRGHAYRDY GHSRRDESYS RGYRNHRSSR ETREYAPPSR GHGYRDYGHS RRHESYSRGY RNHPSSRETR DYAPPHRDYA YRDYGHSSWD EHSSRGYSYH DGYGEALGRD HSEHLSGSSY RDALQRYGTS HGAPPARGPR MSYGGSTCHA YSNTRDRYGR SWESYSSCGD FHYCDREHVC RKDQRNPPSL GRVLPDPREA YGSSSYVASI VDGGESRSEK GDSSRY | |
Greater than 90% as determined by SDS-PAGE. |
159163 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |