missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RBMY1A1 Protein

A DNA sequence encoding the Homo sapiens (Human) RBMY1A1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7035-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16057130

  • 2530.00 € / 1mg

Estimated Shipment: 23-11-2022
to see stock.
Add to basket

Description

Description

Specifications

Specifications

NP_001307873.1
RBMY1A1 Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
378949
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RBMY1A1
Recombinant Protein
E. coli
MVEADHPGKL FIGGLNRETN EKMLKAVFGK HGPISEVLLI KDRTSKSRGF AFITFENPAD AKNAAKDMNG KSLHGKAIKV EQAKKPSFQS GGRRRPPASS RNRSPSGSLR SARGSRGGTR GWLPSHEGHL DDGGYTPDLK MSYSRGLIPV KRGPSSRSGG PPPKKSAPSA VARSNSWMGS QGPMSQRREN YGVPPRRATI SSWRNDRMST RHDGYATNDG NHPSCQETRD YAPPSRGYAY RDNGHSNRDE HSSRGYRNHR SSRETRDYAP PSRGHAYRDY GHSRRDESYS RGYRNRRSSR ETREYAPPSR GHGYRDYGHS RRHESYSRGY RNHPSSRETR DYAPPHRDYA YRDYGHSSWD EHSSRGYSYH DGYGEALGRD HSEHLSGSSY RDALQRYGTS HGAPPARGPR MSYGGSTCHA YSNTRDRYGR SWESYSSCGD FHYCDREHVC RKDQRNPPS
Videos
SDS
Documents
Certificates
Promotions

Promotions