missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RALGPS1 Protein

A DNA sequence encoding the Homo sapiens (Human) RALGPS1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7659-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16090031

  • 493.03 € / 100µg

Estimated Shipment: 12-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001177657.1
RALGPS1 Protein
100 μg
Not determined.
Full Length of Isoform 4
Human
His-SUMO
Tris-based buffer, 50% glycerol
9649
Greater than 90% as determined by SDS-PAGE.
Research Use Only
RALGPS1
Recombinant Protein
E. coli
MYKRNGLMAS VLVTSATPQG SSSSDSLEGQ SCDYASKSYD AVVFDVLKVT PEEFASQITL MDIPVFKAIQ PEELASCGWS KKEKHSLAPN VVAFTRRFNQ VSFWVVREIL TAQTLKIRAE ILSHFVKIAK KLLELNNLHS LMSVVSALQS APIFRLTKTW ALLNRKDKTT FEKLDYLMSK EDNYKRTREY IRSLKMVPSI PYLGIYLLDL IYIDSAYPAS GSIMENEQRS NQMNNILRII ADLQVSCSYD HLTTLPHVQK YLKSVRYIEE LQKFVEDDNY KLSLRIEPGS SSPRLVSSKE DLAAM
Videos
SDS
Documents
Certificates
Promotions

Promotions