missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rab effector Noc2 Protein
A DNA sequence encoding the Homo sapiens (Human) Rab effector Noc2, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001177340.1 |
---|---|
Gene ID (Entrez) | 9501 |
Name | Rab effector Noc2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083541
|
enQuireBio™
QP8248-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003551
|
enQuireBio™
QP8248-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16093541
|
enQuireBio™
QP8248-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16013551
|
enQuireBio™
QP8248-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023551
|
enQuireBio™
QP8248-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033551
|
enQuireBio™
QP8248-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001177340.1 | |
Rab effector Noc2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
9501 | |
Research Use Only | |
RPH3AL | |
Recombinant Protein | |
E. coli | |
MADTIFGSGN DQWVCPNDRQ LALRAKLQTG WSVHTYQTEK QRRKQHLSPA EVEAILQVIQ RAERLDVLEQ QRIGRLVERL ETMRRNVMGN GLSQCLLCGE VLGFLGSSSV FCKDCRKKVC TKCGIEASPG QKRPLWLCKI CSEQREVWKR SGAWFYKGLP KYILPLKTPG RADDPHFRPL PTEPAEREPR SSETSRIYTW ARGRVVSSDS DSDSDLSSSS LEDRLPSTGV RDRKGDKPWK ESGGSVEAPR MGFTHPPGHL SGCQSSLASG ETGTGSADPP GGPRPGLTRR APVKDTPGRA PAADAAPAGP SSCLG | |
Greater than 90% as determined by SDS-PAGE. |