missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PTPRB Protein
A DNA sequence encoding the Homo sapiens (Human) PTPRB, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001103224.1 |
---|---|
Gene ID (Entrez) | 5787 |
Name | PTPRB Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16074120
|
enQuireBio™
QP6556-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094120
|
enQuireBio™
QP6556-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16084120
|
enQuireBio™
QP6556-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16004130
|
enQuireBio™
QP6556-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014130
|
enQuireBio™
QP6556-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024130
|
enQuireBio™
QP6556-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001103224.1 | |
PTPRB Protein | |
Not determined. | |
Cytoplasmic Domain | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5787 | |
Research Use Only | |
PTPRB | |
Recombinant Protein | |
E. coli | |
RQKVSHGRER PSARLSIRRD RPLSVHLNLG QKGNRKTSCP IKINQFEGHF MKLQADSNYL LSKEYEELKD VGRNQSCDIA LLPENRGKNR YNNILPYDAT RVKLSNVDDD PCSDYINASY IPGNNFRREY IVTQGPLPGT KDDFWKMVWE QNVHNIVMVT QCVEKGRVKC DHYWPADQDS LYYGDLILQM LSESVLPEWT IREFKICGEE QLDAHRLIRH FHYTVWPDHG VPETTQSLIQ FVRTVRDYIN RSPGAGPTVV HCSAGVGRTG TFIALDRILQ QLDSKDSVDI YGAVHDLRLH RVHMVQTECQ YVYLHQCVRD VLRARKLRSE QENPLFPIYE NVNPEYHRDP VYSRH | |
Greater than 90% as determined by SDS-PAGE. |