missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Pseudokinase FAM20A Protein

A DNA sequence encoding the Homo sapiens (Human) Pseudokinase FAM20A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9870-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068871

  • 595.74 € / 100µg

Estimated Shipment: 12-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001230675.1
Pseudokinase FAM20A Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
54757
Greater than 90% as determined by SDS-PAGE.
Research Use Only
FAM20A
Recombinant Protein
Yeast
QLRPRERPRG CPCTGRASSL ARDSAAAASD PGTIVHNFSR TEPRTEPAGG SHSGSSSKLQ ALFAHPLYNV PEEPPLLGAE DSLLASQEAL RYYRRKVARW NRRHKMYREQ MNLTSLDPPL QLRLEASWVQ FHLGINRHGL YSRSSPVVSK LLQDMRHFPT ISADYSQDEK ALLGACDCTQ IVKPSGVHLK LVLRFSDFGK AMFKPMRQQR DEETPVDFFY FIDFQRHNAE IAAFHLDRIL DFRRVPPTVG RIVNVTKEIL EVTKNEILQS VFFVSPASNV CFFAKCPYMC KTEYAVCGNP HLLEGSLSAF LPSLNLAPRL SVPNPWIRSY TLAGKEEWEV NPLYCDTVKQ IYPYNNSQRL LNVIDMAIFD FLIGNMDRHH YEMFTKFGDD GFLIHLDNAR GFGRHSHDEI SILSPLSQCC MIKKKTLLHL QLLAQADYRL SDVMRESLLE DQLSPVLTEP HLLALDRRLQ TILRTVEGCI VAHGQQSVIV DGPVEQLAPD SGQANLTS
Videos
SDS
Documents
Certificates
Promotions

Promotions