missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Pseudokinase FAM20A Protein
A DNA sequence encoding the Homo sapiens (Human) Pseudokinase FAM20A, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_001230675.1 |
---|---|
Gene ID (Entrez) | 54757 |
Name | Pseudokinase FAM20A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068871
|
enQuireBio™
QP9870-YE-100UG |
100 μg |
645.00 €
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16008881
|
enQuireBio™
QP9870-YE-500UG |
500 μg |
1740.00 €
500µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078871
|
enQuireBio™
QP9870-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098871
|
enQuireBio™
QP9870-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018881
|
enQuireBio™
QP9870-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16088871
|
enQuireBio™
QP9870-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001230675.1 | |
Pseudokinase FAM20A Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
54757 | |
Research Use Only | |
FAM20A | |
Recombinant Protein | |
Yeast | |
QLRPRERPRG CPCTGRASSL ARDSAAAASD PGTIVHNFSR TEPRTEPAGG SHSGSSSKLQ ALFAHPLYNV PEEPPLLGAE DSLLASQEAL RYYRRKVARW NRRHKMYREQ MNLTSLDPPL QLRLEASWVQ FHLGINRHGL YSRSSPVVSK LLQDMRHFPT ISADYSQDEK ALLGACDCTQ IVKPSGVHLK LVLRFSDFGK AMFKPMRQQR DEETPVDFFY FIDFQRHNAE IAAFHLDRIL DFRRVPPTVG RIVNVTKEIL EVTKNEILQS VFFVSPASNV CFFAKCPYMC KTEYAVCGNP HLLEGSLSAF LPSLNLAPRL SVPNPWIRSY TLAGKEEWEV NPLYCDTVKQ IYPYNNSQRL LNVIDMAIFD FLIGNMDRHH YEMFTKFGDD GFLIHLDNAR GFGRHSHDEI SILSPLSQCC MIKKKTLLHL QLLAQADYRL SDVMRESLLE DQLSPVLTEP HLLALDRRLQ TILRTVEGCI VAHGQQSVIV DGPVEQLAPD SGQANLTS | |
Greater than 90% as determined by SDS-PAGE. |