missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PRRX1 Protein
A cDNA sequence encoding the PRRX1 was constructed and used to recombinantly synthesize the protein.
137.00 € - 7875.00 €
Specifications
Name | PRRX1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15932729
|
enQuireBio™
QP13173-2UG |
2 μg |
137.00 €
2µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15912729
|
enQuireBio™
QP13173-10UG |
10 μg |
212.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15922729
|
enQuireBio™
QP13173-1MG |
1 mg |
7875.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PRRX1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
PRRX1 protein solution (0.25 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 50% glycerol and 2mM DTT. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMTSSYG HVLERQPALG GRLDSPGNLD TLQAKKNFSV SHLLDLEEAG DMVAAQADEN VGEAGRSLLE SPGLTSGSDT PQQDNDQLNS EEKKKRKQRR NRTTFNSSQL QALERVFERT HYPDAFVRED LARRVNLTEA RVQVWFQNRR AKFRRNERAM LANKNASLLK SYSGDVTAVE QPIVPRPAPR PTDYLSWGTA SPYRSSSLPR CCLHEGLHNG F | |
Greater than 85.0% as determined by SDS-PAGE. |