missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Protein yippee-like 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Protein yippee-like 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6897-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096620

  • 530.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001138996.1
Protein yippee-like 3 Protein
Research Use Only
YPEL3
Recombinant Protein
E. coli
MVRISKPKTF QAYLDDCHRR YSCAHCRAHL ANHDDLISKS FQGSQGRAYL FNSVVNVGCG PAEERVLLTG LHAVADIHCE NCKTTLGWKY EQAFESSQKY KEGKYIIELN HMIKDNGWD
Greater than 90% as determined by SDS-PAGE.
83719
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions