missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein SSX2 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein SSX2, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_001157889.1 |
---|---|
Gene ID (Entrez) | 6757 |
Name | Protein SSX2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16099220
|
enQuireBio™
QP7535-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16019230
|
enQuireBio™
QP7535-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16009230
|
enQuireBio™
QP7535-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029230
|
enQuireBio™
QP7535-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039230
|
enQuireBio™
QP7535-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049230
|
enQuireBio™
QP7535-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001157889.1 | |
Protein SSX2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
6757 | |
Research Use Only | |
SSX2 | |
Recombinant Protein | |
E. coli | |
MNGDDAFARR PTVGAQIPEK IQKAFDDIAK YFSKEEWEKM KASEKIFYVY MKRKYEAMTK LGFKATLPPF MCNKRAEDFQ GNDLDNDPNR GNQVERPQMT FGRLQGISPK IMPKKPAEEG NDSEEVPEAS GPQNDGKELC PPGKPTTSEK IHERSGPKRG EHAWTHRLRE RKQLVIYEEI SDPEEDDE | |
Greater than 90% as determined by SDS-PAGE. |