missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Protein BEX3 Protein
A DNA sequence encoding the Homo sapiens (Human) Protein BEX3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6419-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_055195.1 | |
Protein BEX3 Protein | |
Research Use Only | |
NGFRAP1 | |
Recombinant Protein | |
E. coli | |
MANIHQENEE MEQPMQNGEE DRPLGGGEGH QPAGNRRGQA RRLAPNFRWA IPNRQINDGM GGDGDDMEIF MEEMREIRRK LRELQLRNCL RILMGELSNH HDHHDEFCLM P | |
Greater than 90% as determined by SDS-PAGE. |
27018 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |