missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Proteasome subunit beta type-4 Protein

A DNA sequence encoding the Homo sapiens (Human) Proteasome subunit beta type-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8363-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16054231

  • 530.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002787.2
Proteasome subunit beta type-4 Protein
Research Use Only
PSMB4
Recombinant Protein
E. coli
TQNPMVTGTS VLGVKFEGGV VIAADMLGSY GSLARFRNIS RIMRVNNSTM LGASGDYADF QYLKQVLGQM VIDEELLGDG HSYSPRAIHS WLTRAMYSRR SKMNPLWNTM VIGGYADGES FLGYVDMLGV AYEAPSLATG YGAYLAQPLL REVLEKQPVL SQTEARDLVE RCMRVLYYRD ARSYNRFQIA TVTEKGVEIE GPLSTETNWD IAHMISGFE
Greater than 90% as determined by SDS-PAGE.
5692
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions