missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Probable E3 SUMO-protein ligase RNF212 Protein

A DNA sequence encoding the Homo sapiens (Human) Probable E3 SUMO-protein ligase RNF212, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6612-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16084530

  • 770.00 € / 100µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001124506.1
Probable E3 SUMO-protein ligase RNF212 Protein
Research Use Only
RNF212
Recombinant Protein
E. coli
MANWVFCNRC FQPPHRTSCF SLTNCGHVYC DACLGKGKKN ECLICKAPCR TVLLSKHTDA DIQAFFMSID SLCKKYSRET SQILEFQEKH RKRLLAFYRE KISRLEESLR KSVLQIEQLQ SMRSSQQTAF STIKSSVSTK PHGCLLPPHS SAPDRLESME VDLSPSPIRK SEIAAGPARI SMISPPQDGR MGPHLTASFC FIPWLTLSKP PVPGECVISR GSPCFCIDVC PHWLLLLAFS SGRHGELTNS KTLPIYAEVQ RAVLFPFQQA EGTLDTFRTP AVSVVFPLCQ FERKKSF
Greater than 80% as determined by SDS-PAGE.
285498
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions