missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PHLDA2 Protein

A DNA sequence encoding the Homo sapiens (Human) PHLDA2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7664-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16090061

Quantity Price
1 493.03 € / 100µg
6 463.44 € / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_003302.1
PHLDA2 Protein
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
7262
Greater than 80% as determined by SDS-PAGE.
Research Use Only
PHLDA2
Recombinant Protein
E. coli
MKSPDEVLRE GELEKRSDSL FQLWKKKRGV LTSDRLSLFP ASPRARPKEL RFHSILKVDC VERTGKYVYF TIVTTDHKEI DFRCAGESCW NAAIALALID FQNRRALQDF RSRQERTAPA APAEDAVAAA AAAPSEPSEP SRPSPQPKPR TP
Videos
SDS
Documents
Certificates
Promotions

Promotions