missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human OXNAD1 Protein

A DNA sequence encoding the Homo sapiens (Human) OXNAD1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7897-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16091461

Quantity Price
1 493.03 € / 100µg
6 463.44 € / 100µg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_612390.1
OXNAD1 Protein
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
92106
Greater than 90% as determined by SDS-PAGE.
Research Use Only
OXNAD1
Recombinant Protein
E. coli
AIRIEAASLR LTLSTLRHLT LTSIMKSKRK TDHMERTASV LRREIVSAAK VCGAASESPS VKSLRLLVAD QDFSFKAGQW VDFFIPGVSV VGGFSICSSP RLLEQERVIE LAVKYTNHPP ALWVHNTCTL DCEVAVRVGG EFFFDPQPAD ASRNLVLIAG GVGINPLLSI LRHAADLLRE QANKRNGYEI GTIKLFYSAK NTSELLFKKN ILDLVNEFPE KIACSLHVTK QTTQINAELK PYITEGRITE KEIRDHISKE TLFYICGPPP MTDFFSKQLE NNHVPKEHIC FEKWW
Videos
SDS
Documents
Certificates
Promotions

Promotions