missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human OXNAD1 Protein
A DNA sequence encoding the Homo sapiens (Human) OXNAD1, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Accession Number | NP_612390.1 |
---|---|
Gene ID (Entrez) | 92106 |
Name | OXNAD1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16091461
|
enQuireBio™
QP7897-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16011471
|
enQuireBio™
QP7897-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 17-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16001471
|
enQuireBio™
QP7897-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16021471
|
enQuireBio™
QP7897-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16031471
|
enQuireBio™
QP7897-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16041471
|
enQuireBio™
QP7897-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_612390.1 | |
OXNAD1 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
92106 | |
Research Use Only | |
OXNAD1 | |
Recombinant Protein | |
E. coli | |
AIRIEAASLR LTLSTLRHLT LTSIMKSKRK TDHMERTASV LRREIVSAAK VCGAASESPS VKSLRLLVAD QDFSFKAGQW VDFFIPGVSV VGGFSICSSP RLLEQERVIE LAVKYTNHPP ALWVHNTCTL DCEVAVRVGG EFFFDPQPAD ASRNLVLIAG GVGINPLLSI LRHAADLLRE QANKRNGYEI GTIKLFYSAK NTSELLFKKN ILDLVNEFPE KIACSLHVTK QTTQINAELK PYITEGRITE KEIRDHISKE TLFYICGPPP MTDFFSKQLE NNHVPKEHIC FEKWW | |
Greater than 90% as determined by SDS-PAGE. |