missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NUDT2 / Ap4A hydrolase Protein

A DNA sequence encoding the Homo sapiens (Human) NUDT2 / Ap4A hydrolase, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6935-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096760

  • 2385.00 € / 1mg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001152.1
NUDT2 / Ap4A hydrolase Protein
Research Use Only
NUDT2
Recombinant Protein
E. coli
MALRACGLII FRRCLIPKVD NNAIEFLLLQ ASDGIHHWTP PKGHVEPGED DLETALRETQ EEAGIEAGQL TIIEGFKREL NYVARNKPKT VIYWLAEVKD YDVEIRLSHE HQAYRWLGLE EACQLAQFKE MKAALQEGHQ FLCSIEA
Greater than 80% as determined by SDS-PAGE.
318
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions