missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Nucleoside diphosphate kinase, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Nucleoside diphosphate kinase, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6425-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16073220

  • 2248.90 € / 1mg

Estimated Shipment: 12-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001273362.1
Nucleoside diphosphate kinase, mitochondrial Protein
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
4833
Greater than 80% as determined by SDS-PAGE.
Research Use Only
NME4
Recombinant Protein
E. coli
SWTRERTLVA VKPDGVQRRL VGDVIQRFER RGFTLVGMKM LQAPESVLAE HYQDLRRKPF YPALIRYMSS GPVVAMVWEG YNVVRASRAM IGHTDSAEAA PGTIRGDFSV HISRNVIHAS DSVEGAQREI QLWFQSSELV SWADGGQHSS IHPA
Videos
SDS
Documents
Certificates
Promotions

Promotions