Learn More
R&D Systems™ Recombinant Human NT-3 Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
25 μg
missing translation for 'unitSize'
25µg
Beschreibung
The Recombinant Human NT-3 Protein is derived from Sf 21 (baculovirus). The Recombinant Human NT-3 Protein has been validated for the following applications: Bioactivity.
Spezifikation
Spezifikation
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 4908 |
| Molecular Weight (g/mol) | 13.7 kDa |
| Name | NT-3 |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived human NT-3 protein Tyr139-Thr257 (K196R) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | HDNF, MGC129711, Nerve growth factor 2, Neurotrophic factor, neurotrophin 3, neurotrophin-3, NGF2, NGF-2, NT3, NT-3, NTF3 |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Mehr anzeigen |