missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NR2C2 Protein

A DNA sequence encoding the Homo sapiens (Human) NR2C2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8855-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16096841

Quantity Price
1 434.28 € / 100µg
6 408.22 € / 100µg
Estimated Shipment
09-06-2021
Add to basket


Description

Description

Specifications

Specifications

NP_001334271.1
NR2C2 Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
22026
Greater than 90% as determined by SDS-PAGE.
Research Use Only
NR2C2
Recombinant Protein
E. coli
SPSPRIQIIS TDSAVASPQR IQIVTDQQTG QKIQIVTAVD ASGSSKQQFI LTSPDGAGTG KVILASPETS SAKQLIFTTS DNLVPGRIQI VTDSASVERL LGKADVQRPQ VVEYCVVCGD KASGRHYGAV SCEGCKGFFK RSVRKNLTYS CRSSQDCIIN KHHRNRCQFC RLKKCLEMGM KMESVQSERK PFDVQREKPS NCAASTEKIY IRKDLRSPLI ATPTFVADKD GARQTGLLDP GMLVNIQQPL IREDGTVLLA ADSKAETSQG ALGTLANVVT SLANLSESLN NGDASEMQPE DQSASEITRA FDTLAKALNT TDSASPPSLA DGIDASGGGS IHVISRDQST PIIEVEGPLL SDTHVTFKLT MPSPMPEYLN VHYICESASR LLFLSMHWAR SIPAFQALGQ DCNTSLVRAC WNELFTLGLA QCAQVMSLST ILAAIVNHLQ NSIQEDKLSG DRIKQVMEHI WKLQEFCNSM AKLDIDGYEY AYLKAIVLFS PDHPGLTGTS QIEKFQEKAQ MELQDYVQKT YSEDTYRLAR ILVRLPALRL MSSNITEELF FTGLIGNVSI DSIIPYILKM ETAEYNGQIT
Videos
SDS
Documents