missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NR1I3 Protein

A DNA sequence encoding the Homo sapiens (Human) NR1I3, was expressed in the hosts and tags indicated.

160.00 € - 2385.00 €

Specifications

Accession Number NP_001070937.1
Gene ID (Entrez) 9970
Name NR1I3 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16049660
View Documents Promotion Details
enQuireBio™
QP7599-EC-100UG
100 μg
530.00 €
100µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16069660
View Documents Promotion Details
enQuireBio™
QP7599-EC-1MG
1 mg
2385.00 €
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16059660
View Documents Promotion Details
enQuireBio™
QP7599-EC-10UG
10 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16079660
View Documents Promotion Details
enQuireBio™
QP7599-EC-200UG
200 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16089660
View Documents Promotion Details
enQuireBio™
QP7599-EC-500UG
500 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
16099660
View Documents Promotion Details
enQuireBio™
QP7599-EC-50UG
50 μg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

Specifications

Specifications

NP_001070937.1
NR1I3 Protein
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
9970
Research Use Only
NR1I3
Recombinant Protein
E. coli
ASREDELRNC VVCGDQATGY HFNALTCEGC KGFFRRTVSK SIGPTCPFAG SCEVSKTQRR HCPACRLQKC LDAGMRKDMI LSAEALALRR AKQAQRRAQQ TPVQLSKEQE ELIRTLLGAH TRHMGTMFEQ FVQFRPPAHL FIHHQPLPTL APVLPLVTHF ADINTFMVLQ VIKFTKDLPV FRSLPIEDQI SLLKGAAVEI CHIVLNTTFC LQTQNFLCGP LRYTIEDGAR VSPTVGFQVE FLELLFHFHG TLRKLQLQEP EYVLLAAMAL FSPDRPGVTQ RDEIDQLQEE MALTLQSYIK GQQRRPRDRF LYAKLLGLLA ELRSINEAYG YQIQHIQGLS AMMPLLQEIC
Greater than 90% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications