missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NKG2-C type II integral membrane Protein

A DNA sequence encoding the Homo sapiens (Human) NKG2-C type II integral membrane protein, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6270-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16072080

Quantity Price
1 2248.90 € / 1mg
3 2115.77 € / 1mg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_002251.2
NKG2-C type II integral membrane Protein
1 mg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
3822
Greater than 90% as determined by SDS-PAGE.
Research Use Only
KLRC2
Recombinant Protein
E. coli
IPFLEQNNSS PNTRTQKARH CGHCPEEWIT YSNSCYYIGK ERRTWEESLL ACTSKNSSLL SIDNEEEMKF LASILPSSWI GVFRNSSHHP WVTINGLAFK HKIKDSDNAE LNCAVLQVNR LKSAQCGSSM IYHCKHKL
Videos
SDS
Documents
Certificates
Promotions

Promotions