missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFA12 Protein

A DNA sequence encoding the Homo sapiens (Human) NDUFA12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8187-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093141

  • 530.00 € / 100µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001245267.1
NDUFA12 Protein
Research Use Only
NDUFA12
Recombinant Protein
E. coli
MELVQVLKRG LQQITGHGGL RGYLRVFFRT NDAKVGTLVG EDKYGNKYYE DNKQFFGRHR WVVYTTEMNG KNTFWDVDGS MVPPEWHRWL HSMTDDPPTT KPLTARKFIW TNHKFNVTGT PEQYVPYSTT RKKIQEWIPP STPYK
Greater than 80% as determined by SDS-PAGE.
55967
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions