missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin-binding protein C, fast-type Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin-binding protein C, fast-type, was expressed in the hosts and tags indicated.
202.00 € - 2155.00 €
Specifications
Accession Number | NP_004524.3 |
---|---|
Gene ID (Entrez) | 4606 |
Name | Myosin-binding protein C, fast-type Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16069400
|
enQuireBio™
QP7562-YE-100UG |
100 μg |
645.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16009410
|
enQuireBio™
QP7562-YE-500UG |
500 μg |
1740.00 €
500µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16079400
|
enQuireBio™
QP7562-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16099400
|
enQuireBio™
QP7562-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019410
|
enQuireBio™
QP7562-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16089400
|
enQuireBio™
QP7562-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004524.3 | |
Myosin-binding protein C, fast-type Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4606 | |
Research Use Only | |
MYBPC2 | |
Recombinant Protein | |
Yeast | |
EPLHLIVEDV TDTTTTLKWR PPNRIGAGGI DGYLVEYCLE GSEEWVPANT EPVERCGFTV KNLPTGARIL FRVVGVNIAG RSEPATLAQP VTIREIAEPP KIRLPRHLRQ TYIRKVGEQL NLVVPFQGKP RPQVVWTKGG APLDTSRVHV RTSDFDTVFF VRQAARSDSG EYELSVQIEN MKDTATIRIR VVEKAGPPIN VMVKEVWGTN ALVEWQAPKD DGNSEIMGYF VQKADKKTME WFNVYERNRH TSCTVSDLIV GNEYYFRVYT ENICGLSDSP GVSKNTARIL KTGITFKPFE YKEHDFRMAP KFLTPLIDRV VVAGYSAALN CAVRGHPKPK VVWMKNKMEI REDPKFLITN YQGVLTLNIR RPSPFDAGTY TCRAVNELGE ALAECKLEVR VPQ | |
Greater than 90% as determined by SDS-PAGE. |