missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Microfibrillar-associated protein 1

A DNA sequence encoding the Homo sapiens (Human) Microfibrillar-associated protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7607-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16089730

  • 2248.90 € / 1mg

Estimated Shipment: 10-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_001284638.1
Microfibrillar-associated protein 1
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
8076
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MFAP5
Recombinant Protein
E. coli
IPLGVNSQRG DDVTQATPET FTEDPNLVND PATDETVLAV LADIAPSTDD LASLSEKNTT AECWDEKFTC TRLYSVHRPV KQCIHQLCFT SLRRMYIVNK EICSRLVCKE HEAMKDELCR QMAGLPPRRL RRSNYFRLPP CENVDLQRPN GL
Videos
SDS
Documents
Certificates
Promotions

Promotions