missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Metallothionein-1E Protein

A DNA sequence encoding the Homo sapiens (Human) Metallothionein-1E, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8895-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16097041

  • 2385.00 € / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_783316.2
Metallothionein-1E Protein
Research Use Only
MT1E
Recombinant Protein
E. coli
NCSCATGGSC TCAGSCKCKE CKCTSCKKSC CSCCPVGCAK CAQGCVCKGA SEKCSC
Greater than 90% as determined by SDS-PAGE.
4493
1 mg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions