missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Major surface glycoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Major surface glycoprotein G, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7412-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
Major surface glycoprotein 1 | |
Research Use Only | |
G | |
Recombinant Protein | |
E. coli | |
HKVTPTTAII QDATSQIKNT TPTYLTQNPQ LGISPSNPSE ITSQITTILA STTPGVKSTL QSTTVKTKNT TTTQTQPSKP TTKQRQNKPP SKPNNDFHFE VFNFVPCSIC SNNPTCWAIC KRIPNKKPGK KTTTKPTKKP TLKTTKKDPK PQTTKSKEVP TTKPTEEPTI NTTKTNIITT LLTSNTTGNP ELTSQMETFH STSSEGNPSP SQVSTTSEYP SQPSSPPNTP RQ | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |