missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Major surface glycoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Major surface glycoprotein G, was expressed in the hosts and tags indicated.
160.00 € - 2385.00 €
Specifications
Name | Major surface glycoprotein 1 |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Gene Symbol | G |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16068740
|
enQuireBio™
QP7412-EC-100UG |
100 μg |
530.00 €
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088740
|
enQuireBio™
QP7412-EC-1MG |
1 mg |
2385.00 €
1mg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16078740
|
enQuireBio™
QP7412-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098740
|
enQuireBio™
QP7412-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16008750
|
enQuireBio™
QP7412-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16018750
|
enQuireBio™
QP7412-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
Major surface glycoprotein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Research Use Only | |
G | |
Recombinant Protein | |
E. coli | |
HKVTPTTAII QDATSQIKNT TPTYLTQNPQ LGISPSNPSE ITSQITTILA STTPGVKSTL QSTTVKTKNT TTTQTQPSKP TTKQRQNKPP SKPNNDFHFE VFNFVPCSIC SNNPTCWAIC KRIPNKKPGK KTTTKPTKKP TLKTTKKDPK PQTTKSKEVP TTKPTEEPTI NTTKTNIITT LLTSNTTGNP ELTSQMETFH STSSEGNPSP SQVSTTSEYP SQPSSPPNTP RQ | |
Greater than 90% as determined by SDS-PAGE. |