missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lymphocyte antigen 6E Protein

A DNA sequence encoding the Homo sapiens (Human) Lymphocyte antigen 6E, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7565-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16069430

  • 530.00 € / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001120685.1
Lymphocyte antigen 6E Protein
Research Use Only
LY6E
Recombinant Protein
E. coli
LMCFSCLNQK SNLYCLKPTI CSDQDNYCVT VSASAGIGNL VTFGHSLSKT CSPACPIPEG VNVGVASMGI SCCQSFLCNF S
Greater than 90% as determined by SDS-PAGE.
4061
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions