Learn More
enQuireBio™ Recombinant Human La-related protein 1B Protein
A DNA sequence encoding the Homo sapiens (Human) La-related protein 1B, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7726-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16070441
Quantity Price
1 493.03 € / 100µg
6 463.44 € / 100µg
Estimated Shipment: 17-01-2022
Add to basket
Specifications
NP_001265533.1
La-related protein 1B Protein
100 μg
Not determined.
Full Length of Isoform 7
Human
His-SUMO
Tris-based buffer, 50% glycerol
55132
Greater than 90% as determined by SDS-PAGE.
Research Use Only
LARP1B
Recombinant Protein
E. coli
MDSRDRGPGT SSVSTSNASP SEGAPLAGSY GCTPHSFPKF QHPSHELLKE NGFTQQVYHK YRRRCLSERK RLGIGQSQEM NTLFRFWSFF LRDHFNKKMY EEFRQLAWED AKENYRYGLE CLFRFYSYGL EKKFRREIFQ DFQEETKKDY ESGQLYGLEK FWAYLKYSQS KTQSIDPKLQ EYLCSFKRLE DFRVDEADPI E