missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Kinesin light chain 3 Protein

A DNA sequence encoding the Homo sapiens (Human) Kinesin light chain 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6265-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16092030

Quantity Price
1 3211.16 € / 1mg
3 3021.38 € / 1mg
Estimated Shipment: 18-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_803136.2
Kinesin light chain 3 Protein
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
147700
Greater than 90% as determined by SDS-PAGE.
Research Use Only
KLC3
Recombinant Protein
E. coli
MSVQVAAPGS AGLGPERLSP EELVRQTRQV VQGLEALRAE HHGLAGHLAE ALAGQGPAAG LEMLEEKQQV VSHSLEAIEL GLGEAQVLLA LSAHVGALEA EKQRLRSQAR RLAQENVWLR EELEETQRRL RASEESVAQL EEEKRHLEFL GQLRQYDPPA ESQQSESPPR RDSLASLFPS EEEERKGPEA AGAAAAQQGG YEIPARLRTL HNLVIQYAGQ GRYEVAVPLC RQALEDLERS SGHCHPDVAT MLNILALVYR DQNKYKEATD LLHDALQIRE QTLGPEHPAV AATLNNLAVL YGKRGRYREA EPLCQRALEI REKVLGADHP DVAKQLNNLA LLCQNQGKFE DVERHYARAL SIYEALGGPH DPNVAKTKNN LASAYLKQNK YQQAEELYKE ILHKEDLPAP LGAPNTGTAG DAEQALRRSS SLSKIRESIR RGSEKLVSRL RGEAAAGAAG MKRAMSLNTL NVDAPRAPGT QFPSWHLDKA PRTLSASTQD LSPH
Videos
SDS
Documents
Certificates
Promotions

Promotions