missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Interleukin enhancer-binding factor 2 Protein

A DNA sequence encoding the Homo sapiens (Human) Interleukin enhancer-binding factor 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8309-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16093921

  • 530.00 € / 100µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001254738.1
Interleukin enhancer-binding factor 2 Protein
Research Use Only
ILF2
Recombinant Protein
E. coli
MRGDRGRGRG GRFGSRGGPG GGFRPFVPHI PFDFYLCEMA FPRVKPAPDE TSFSEALLKR NQDLAPNSAE QASILSLVTK INNVIDNLIV APGTFEVQIE EVRQVGSYKK GTMTTGHNVA DLVVILKILP TLEAVAALGN KVVESLRAQD PSEVLTMLTN ETGFEISSSD ATVKILITTV PPNLRKLDPE LHLDIKVLQS ALAAIRHARW FEENASQSTV KVLIRLLKDL RIRFPGFEPL TPWILDLLGH YAVMNNPTRQ PLALNVAYRR CLQILAAGLF LPGSVGITDP CESGNFRVHT VMTLEQQDMV CYTAQTLVRI LSHGGFRKIL GQEGDASYLA SEISTWDGVI VTPSEKAYEK PPEKKEGEEE EENTEEPPQG EEEESMETQE
Greater than 90% as determined by SDS-PAGE.
3608
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions