missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Interleukin-26 Protein

A DNA sequence encoding the Homo sapiens (Human) Interleukin-26, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9904-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16068961

Quantity Price
1 595.74 € / 100µg
5 560.65 € / 100µg
Estimated Shipment: 17-01-2022

Add to basket


Description

Description

Specifications

Specifications

NP_060872.1
Interleukin-26 Protein
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
55801
Greater than 90% as determined by SDS-PAGE.
Research Use Only
IL26
Recombinant Protein
Yeast
KHKQSSFTKS CYPRGTLSQA VDALYIKAAW LKATIPEDRI KNIRLLKKKT KKQFMKNCQF QEQLLSFFME DVFGQLQLQG CKKIRFVEDF HSLRQKLSHC ISCASSAREM KSITRMKRIF YRIGNKGIYK AISELDILLS WIKKLLESSQ
Videos
SDS
Documents
Certificates
Promotions

Promotions