missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Integrin alpha-IIb Protein

A DNA sequence encoding the Homo sapiens (Human) Integrin alpha-IIb, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6239-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16091830

  • 493.03 € / 100µg

Estimated Shipment: 12-01-2022
Add to basket

Description

Description

Specifications

Specifications

NP_000410.2
Integrin alpha-IIb Protein
100 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
3674
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ITGA2B
Recombinant Protein
E. coli
CVPQLQLTAS VTGSPLLVGA DNVLELQMDA ANEGEGAYEA ELAVHLPQGA HYMRALSNVE GFERLICNQK KENETRVVLC ELGNPMKKNA QIGIAMLVSV GNLEEAGESV SFQLQIRSKN SQNPNSKIVL LDVPVRAEAQ VELRGNSFPA SLVVAAEEGE REQNSLDSWG PKVEHTYELH NNGPGTVNGL HLSIHLPGQS QPSDLLYILD IQPQGGLQCF PQPPVNPLKV DWGLPIPSPS PIHPAHHKR
Videos
SDS
Documents
Certificates
Promotions

Promotions